Order Entry
Slovakia
ContactUsLinkComponent
Biotinylated Human ROR1, His,Avitag (RO1-H82E6) is expressed from human 293 cells (HEK293). It contains AA Gln 30 - Glu 403 (Accessi on Q01973-1). 1 * 25 µG
Catalog # ACRBRO1-H82E6-25UG
Supplier:  ACRO Biosystems
Biotinylated Human ROR1, His,Avitag (RO1-H82E6) is expressed from human 293 cells (HEK293). It contains AA Gln 30 - Glu 403 (Accessi on Q01973-1). 1 * 25 µG
Catalog # ACRBRO1-H82E6-25UG
Supplier:  ACRO Biosystems
Frequently Bought Together