Order Entry
Slovakia
ContactUsLinkComponent
Human Her2, Tag Free (HE2-H5212) is expressed from human 293 cells (HEK293). It contains AA Thr 23 - Thr 652 (Accession P04626-1). 1 * 500 µG
Catalog # ACRBHE2-H5212500UG
Supplier:  ACRO Biosystems
Human Her2, Tag Free (HE2-H5212) is expressed from human 293 cells (HEK293). It contains AA Thr 23 - Thr 652 (Accession P04626-1). 1 * 500 µG
Catalog # ACRBHE2-H5212500UG
Supplier:  ACRO Biosystems
Frequently Bought Together