Order Entry
Slovakia
ContactUsLinkComponent
Mouse monoclonal antibody is produced from a hybridoma resulting from fusion of SP20 myeloma and B-lymphocytes obtained from a mouse immunized with Bevacizumab F(ab)sub2sub. 1 * 200 µG
Catalog # ACRBBEB-BY13-200UG
Supplier:  ACRO Biosystems
Mouse monoclonal antibody is produced from a hybridoma resulting from fusion of SP20 myeloma and B-lymphocytes obtained from a mouse immunized with Bevacizumab F(ab)sub2sub. 1 * 200 µG
Catalog # ACRBBEB-BY13-200UG
Supplier:  ACRO Biosystems
Frequently Bought Together