Order Entry
Slovakia
ContactUsLinkComponent
Recombinat Protein,Biotinylated Human BLAME SLAMF8 Protein, His,Avitag™,Biotinylated Human BLAME, His,Avitag (BLE-H82E4) is expresse d from human 293 cells (HEK293). It contains AA Ala 23 - Asp 233 (Accession Q9P0V8-1).This protein carries a polyhist 1 * 200 µG
Catalog # ACRBBLE-H82E4200UG
Supplier:  ACRO Biosystems
Recombinat Protein,Biotinylated Human BLAME SLAMF8 Protein, His,Avitag™,Biotinylated Human BLAME, His,Avitag (BLE-H82E4) is expresse d from human 293 cells (HEK293). It contains AA Ala 23 - Asp 233 (Accession Q9P0V8-1).This protein carries a polyhist 1 * 200 µG
Catalog # ACRBBLE-H82E4200UG
Supplier:  ACRO Biosystems
Frequently Bought Together