Order Entry
Slovakia
ContactUsLinkComponent
Human EGF, Mouse IgG2a Fc Tag (EGF-H525b) is expressed from human 293 cells (HEK293). It contains AA Asn 971 - Arg 1023 (Accession P 01133-1). 1 * 100 µG
Catalog # ACRBEGF-H525B100UG
Supplier:  ACRO Biosystems
Human EGF, Mouse IgG2a Fc Tag (EGF-H525b) is expressed from human 293 cells (HEK293). It contains AA Asn 971 - Arg 1023 (Accession P 01133-1). 1 * 100 µG
Catalog # ACRBEGF-H525B100UG
Supplier:  ACRO Biosystems
Frequently Bought Together