Recombinat Protein,Biotinylated Human LOXL2 Protein, His,Avitag™,Biotinylated Human LOXL2, His,Avitag (LO2-H82E3) is expressed from human 293 cells (HEK293). It contains AA Gln 26 - Gln 774 (Accession Q9Y4K0-1).This protein carries a polyhistidine ta 1 * 200 µG
Catalog # ACRBLO2-H82E3200UG
Supplier: ACRO Biosystems
Frequently Bought Together