Order Entry
Slovakia
ContactUsLinkComponent
Recombinat Protein,Human Transthyretin Prealbumin Protein, His Tag,Human Transthyretin, His Tag (TTR-H5223) is expressed from human 293 cells (HEK293). It contains AA Gly 21 - Glu 147 (Accession NP000362).This protein carries a polyhistidine tag at t 1 * 1 mg
Catalog # ACRBTTR-H5223-1MG
Supplier:  ACRO Biosystems
Recombinat Protein,Human Transthyretin Prealbumin Protein, His Tag,Human Transthyretin, His Tag (TTR-H5223) is expressed from human 293 cells (HEK293). It contains AA Gly 21 - Glu 147 (Accession NP000362).This protein carries a polyhistidine tag at t 1 * 1 mg
Catalog # ACRBTTR-H5223-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together