Order Entry
Slovakia
ContactUsLinkComponent
Cynomolgus Siglec-15, His Tag (SG5-C52H6) is expressed from human 293 cells (HEK293). It contains AA Phe 20 - Thr 263 (Accession A0A 2K5UY47-1). 1 * 100 µG
Catalog # ACRBSG5-C52H6100UG
Supplier:  ACRO Biosystems
Cynomolgus Siglec-15, His Tag (SG5-C52H6) is expressed from human 293 cells (HEK293). It contains AA Phe 20 - Thr 263 (Accession A0A 2K5UY47-1). 1 * 100 µG
Catalog # ACRBSG5-C52H6100UG
Supplier:  ACRO Biosystems
Frequently Bought Together