Zadaní objednávky
Slovakia
ContactUsLinkComponent
Human IFN-gamma R1, Fc Tag (IF1-H5254) is expressed from human 293 cells (HEK293). It contains AA Glu 18 - Gly 245 (Accession AAH053 33). 1 * 1 mg
Catalog # ACRBIF1-H5254-1MG
Supplier:  ACRO Biosystems
Human IFN-gamma R1, Fc Tag (IF1-H5254) is expressed from human 293 cells (HEK293). It contains AA Glu 18 - Gly 245 (Accession AAH053 33). 1 * 1 mg
Catalog # ACRBIF1-H5254-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together