Order Entry
Slovakia
ContactUsLinkComponent
Human CD20 Full Length, His Tag (HEK293) (SPR verified) (CD0-H52H3) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Pro 297 (Accession P11836-1). 1 * 500 µG
Catalog # ACRBCD0-H52H3500UG
Supplier:  ACRO Biosystems
Human CD20 Full Length, His Tag (HEK293) (SPR verified) (CD0-H52H3) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Pro 297 (Accession P11836-1). 1 * 500 µG
Catalog # ACRBCD0-H52H3500UG
Supplier:  ACRO Biosystems
Frequently Bought Together