Order Entry
Slovakia
ContactUsLinkComponent
Rhesus macaque PLGF, His Tag (PGF-R52H0) is expressed from human 293 cells (HEK293). It contains AA Leu 19 - Arg 170 (Accession F7HB 10-1). 1 * 1 mg
Catalog # ACRBPGF-R52H0-1MG
Supplier:  ACRO Biosystems
Rhesus macaque PLGF, His Tag (PGF-R52H0) is expressed from human 293 cells (HEK293). It contains AA Leu 19 - Arg 170 (Accession F7HB 10-1). 1 * 1 mg
Catalog # ACRBPGF-R52H0-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together