Order Entry
Slovakia
ContactUsLinkComponent
Mouse CTLA-4, Fc Tag (CT4-M5256) is expressed from human 293 cells (HEK293). It contains AA Glu 36 - Phe 162 (Accession NP033973). 1 * 1 mg
Catalog # ACRBCT4-M5256-1MG
Supplier:  ACRO Biosystems
Mouse CTLA-4, Fc Tag (CT4-M5256) is expressed from human 293 cells (HEK293). It contains AA Glu 36 - Phe 162 (Accession NP033973). 1 * 1 mg
Catalog # ACRBCT4-M5256-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together