Order Entry
Slovakia
ContactUsLinkComponent
Human CTGF, His Tag (CTF-H52H5) is expressed from human 293 cells (HEK293). It contains AA Gln 27 - Ala 349 (Accession Q5M8T4-1). 1 * 100 µG
Catalog # ACRBCTF-H52H5100UG
Supplier:  ACRO Biosystems
Human CTGF, His Tag (CTF-H52H5) is expressed from human 293 cells (HEK293). It contains AA Gln 27 - Ala 349 (Accession Q5M8T4-1). 1 * 100 µG
Catalog # ACRBCTF-H52H5100UG
Supplier:  ACRO Biosystems
Frequently Bought Together