Order Entry
Slovakia
ContactUsLinkComponent
Canine CD47, His Tag (CD7-C52H5) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Asn 140 (Accession F1P6D7-1). 1 * 1 mg
Catalog # ACRBCD7-C52H5-1MG
Supplier:  ACRO Biosystems
Canine CD47, His Tag (CD7-C52H5) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Asn 140 (Accession F1P6D7-1). 1 * 1 mg
Catalog # ACRBCD7-C52H5-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together