Order Entry
Slovakia
ContactUsLinkComponent
Human Nectin-1, His Tag (PV1-H5223) is expressed from human 293 cells (HEK293). It contains AA Gln 31 - Thr 334 (Accession NP002846) . 1 * 100 µG
Catalog # ACRBPV1-H5223100UG
Supplier:  ACRO Biosystems
Human Nectin-1, His Tag (PV1-H5223) is expressed from human 293 cells (HEK293). It contains AA Gln 31 - Thr 334 (Accession NP002846) . 1 * 100 µG
Catalog # ACRBPV1-H5223100UG
Supplier:  ACRO Biosystems
Frequently Bought Together