Order Entry
Slovakia
ContactUsLinkComponent
Mouse CD47, Mouse IgG2a Fc Tag, low endotoxin (CD7-M525a) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Lys 14 0 (Accession Q61735-1). 1 * 1 mg
Catalog # ACRBCD7-M525A-1MG
Supplier:  ACRO Biosystems
Mouse CD47, Mouse IgG2a Fc Tag, low endotoxin (CD7-M525a) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Lys 14 0 (Accession Q61735-1). 1 * 1 mg
Catalog # ACRBCD7-M525A-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together