Order Entry
Slovakia
ContactUsLinkComponent
Recombinat Protein,Rabbit PD-1 PDCD1 Protein, His Tag,Rabbit PD-1, His Tag (PD1-R52H0) is expressed from human 293 cells (HEK293). I t contains AA Leu 25 - Gln 167 (Accession G1SUF0).This protein carries a polyhistidine tag at the C-terminus. The pro 1 * 1 mg
Catalog # ACRBPD1-R52H0-1MG
Supplier:  ACRO Biosystems
Recombinat Protein,Rabbit PD-1 PDCD1 Protein, His Tag,Rabbit PD-1, His Tag (PD1-R52H0) is expressed from human 293 cells (HEK293). I t contains AA Leu 25 - Gln 167 (Accession G1SUF0).This protein carries a polyhistidine tag at the C-terminus. The pro 1 * 1 mg
Catalog # ACRBPD1-R52H0-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together