Order Entry
Slovakia
ContactUsLinkComponent
Human HMGB1, Mouse IgG2a Fc Tag, low endotoxin (HM1-H5255) is expressed from human 293 cells (HEK293). It contains AA Gly 2 - Glu 21 5 (Accession P09429-1). 1 * 1 mg
Catalog # ACRBHM1-H5255-1MG
Supplier:  ACRO Biosystems
Human HMGB1, Mouse IgG2a Fc Tag, low endotoxin (HM1-H5255) is expressed from human 293 cells (HEK293). It contains AA Gly 2 - Glu 21 5 (Accession P09429-1). 1 * 1 mg
Catalog # ACRBHM1-H5255-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together