Human CD19 (20-291), Fc Tag, low endotoxin (Super affinity) (CD9-H5251) is expressed from human 293 cells (HEK293). It contains AA P ro 20 - Lys 291 (Accession P15391-1). 1 * 1 mg
Catalog # ACRBCD9-H5251-1MG
Supplier: ACRO Biosystems
Frequently Bought Together