Order Entry
Slovakia
ContactUsLinkComponent
Human VEGF R2, Strep Tag (KDR-H5280) is expressed from human 293 cells (HEK293). It contains AA Ala 20 - Glu 764 (Accession AAI31823 ). 1 * 25 µG
Catalog # ACRBKDR-H5280-25UG
Supplier:  ACRO Biosystems
Human VEGF R2, Strep Tag (KDR-H5280) is expressed from human 293 cells (HEK293). It contains AA Ala 20 - Glu 764 (Accession AAI31823 ). 1 * 25 µG
Catalog # ACRBKDR-H5280-25UG
Supplier:  ACRO Biosystems
Frequently Bought Together