Order Entry
Slovakia
ContactUsLinkComponent
Biotinylated Cynomolgus CD3ECD3D Heterodimer Protein, His,AvitagTag Free (CDD-C82W6) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Asp 117 (CD3E) Phe 22 - Ala 105 (CD3D) (Accession Q95LI5-1 (CD3E) Q95LI8-1 (CD3D)). 1 * 200 µG
Catalog # ACRBCDD-C82W6200UG
Supplier:  ACRO Biosystems
Biotinylated Cynomolgus CD3ECD3D Heterodimer Protein, His,AvitagTag Free (CDD-C82W6) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Asp 117 (CD3E) Phe 22 - Ala 105 (CD3D) (Accession Q95LI5-1 (CD3E) Q95LI8-1 (CD3D)). 1 * 200 µG
Catalog # ACRBCDD-C82W6200UG
Supplier:  ACRO Biosystems
Frequently Bought Together