Order Entry
Slovakia
ContactUsLinkComponent
Mouse PD-1, Fc Tag (PD1-M5259) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Gln 167 (Accession NP032824). 1 * 1 mg
Katalógové číslo ACRBPD1-M5259-1MG
Supplier:  ACRO Biosystems
Mouse PD-1, Fc Tag (PD1-M5259) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Gln 167 (Accession NP032824). 1 * 1 mg
Katalógové číslo ACRBPD1-M5259-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together