Order Entry
Slovakia
ContactUsLinkComponent
Human CD133, His Tag (CD3-H52H4) is expressed from human 293 cells (HEK293). It contains AA Gly 20 - His 865 (Accession O43490-1). 1 * 100 µG
Catalog # ACRBCD3-H52H4100UG
Supplier:  ACRO Biosystems
Human CD133, His Tag (CD3-H52H4) is expressed from human 293 cells (HEK293). It contains AA Gly 20 - His 865 (Accession O43490-1). 1 * 100 µG
Catalog # ACRBCD3-H52H4100UG
Supplier:  ACRO Biosystems
Frequently Bought Together