Terpene Mixture 2500µg/mL in Hexane 1 * 1 mL
Catalog # EHERGA09000634HE
Supplier: EHRENSTORFER
Frequently Bought Together
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
VWR®, Conductivity standards at 25 °C, potassium chloride solutions
Supplier: VWR Chemicals
Short-term tubes for accuro®
Supplier: Dräger
Silver nitrate ≥99%
Supplier: Thermo Fisher Scientific