Order Entry
Slovakia
ContactUsLinkComponent
Overflow pipettes n. Daffert with glass patent tap, full discharge to the burette tip n 0278-15-177 1 * 1 items
Catalog # NEUB0278-15-177
Overflow pipettes n. Daffert with glass patent tap, full discharge to the burette tip n 0278-15-177 1 * 1 items
Catalog # NEUB0278-15-177
Frequently Bought Together