Order Entry
Slovakia
ContactUsLinkComponent
TWEEN® 80 (POLYOXYETHYLENESORBITAN MONOLAURATE), 10% AQUEOUS SOLUTION (PROTEOMIC GRADE) CRIMP STYLE VIAL TWEEN® 80 IS A NON-IONIC D ETERGENT THAT IS PROVIDED FOR RESEARCH PURPOSES OR AS A 10% PROTEOMIC GRADE SOLUTION. 1 * 50 mL
Catalog # GENODG513C
Supplier:  G BIOSCIENCES
TWEEN® 80 (POLYOXYETHYLENESORBITAN MONOLAURATE), 10% AQUEOUS SOLUTION (PROTEOMIC GRADE) CRIMP STYLE VIAL TWEEN® 80 IS A NON-IONIC D ETERGENT THAT IS PROVIDED FOR RESEARCH PURPOSES OR AS A 10% PROTEOMIC GRADE SOLUTION. 1 * 50 mL
Catalog # GENODG513C
Supplier:  G BIOSCIENCES
Frequently Bought Together