Bottle head assembly, for 1260/1290 Infinity II Preperative LC system, includes bottle cap, tubing PTFE-ESD 2 m, fittings, and solvent inlet filter. 1 * 1 items
Catalog # HEWLG7161-60042
Supplier: AGILENT HEWLETT PACKARD
Frequently Bought Together