Order Entry
Slovakia
ContactUsLinkComponent
Counting chambers, Helber (Thoma with special depth), depth 0.02 mm, double net ruling, dark line, without clamps, with two cover glasses of 0.4 mm thickness, CE marked 1 * 1 items
Catalog # MARI0642710-1
Supplier:  MARIENFELD
Counting chambers, Helber (Thoma with special depth), depth 0.02 mm, double net ruling, dark line, without clamps, with two cover glasses of 0.4 mm thickness, CE marked 1 * 1 items
Catalog # MARI0642710-1
Supplier:  MARIENFELD
Frequently Bought Together