4.5ML SKIRTED CRYOGENIC VIAL, INTERNAL THREAD, SILICONE SEAL IN CAP (STERILE) NATURA
STARE3110-6142
: STARLAB
4.5ML SKIRTED CRYOGENIC VIAL, INTERNAL THREAD, SILICONE SEAL IN CAP (STERILE) NATURA
STARE3110-6142
: STARLAB
Frequently Bought Together
Microliter™ 700 series syringes
: HAMILTON BONADUZ
VWR®, Screw Caps for Screw Neck Vials ND24
: VWR Collection
VWR®, Short Thread Vials ND9
: VWR Collection
VWR®, Screw Caps for Short Thread Vials ND9
: VWR Collection
Flat bottom chambers
: CAMAG
VWR®, Snap Cap Vials
: VWR Collection
Exhaust filters, V3.0
: S.C.A.T.
VWR®, Crimp Neck Vials ND11
: VWR Collection
VWR®, Crimp Neck Headspace Vials ND20
: VWR Collection
Florisil® 60-100 mesh for chromatography
: VWR Chemicals
Air valves for safetycaps generation 2.0
: S.C.A.T.
Inlet liners
: AGILENT HEWLETT PACKARD