Order Entry
Slovakia
ContactUsLinkComponent
An ELISA kit for the detection of FGF1 (Human). This uses Sandwich ELISA, Double Antibody and has a sensitivity of 18.75pg/ml. 1 * 1 KIT
Catalog # GENOIT1846
Supplier:  G BIOSCIENCES
An ELISA kit for the detection of FGF1 (Human). This uses Sandwich ELISA, Double Antibody and has a sensitivity of 18.75pg/ml. 1 * 1 KIT
Catalog # GENOIT1846
Supplier:  G BIOSCIENCES
Frequently Bought Together