Order Entry
Slovakia
ContactUsLinkComponent
An ELISA kit for the detection of Pepsinogen (Camel). This uses Sandwich ELISA, Double Antibody and has a sensitivity of 3.75ng/ml. 1 * 1 KIT
Catalog # GENOIT1225
Supplier:  G BIOSCIENCES
An ELISA kit for the detection of Pepsinogen (Camel). This uses Sandwich ELISA, Double Antibody and has a sensitivity of 3.75ng/ml. 1 * 1 KIT
Catalog # GENOIT1225
Supplier:  G BIOSCIENCES
Frequently Bought Together