Order Entry
Slovakia
ContactUsLinkComponent
An ELISA kit for the detection of COR (Bovine). This uses Competitive ELISA, Coated with Antigen and has a sensitivity of 0.234ng/ml . 1 * 1 KIT
Catalog # GENOIT1193
Supplier:  G BIOSCIENCES
An ELISA kit for the detection of COR (Bovine). This uses Competitive ELISA, Coated with Antigen and has a sensitivity of 0.234ng/ml . 1 * 1 KIT
Catalog # GENOIT1193
Supplier:  G BIOSCIENCES
Frequently Bought Together