Order Entry
Slovakia
ContactUsLinkComponent
Our No-Waste format adjuvant minimizes waste and risk of cross contamination. Small dose packaging allows researchers to assign a vial per project or animal without concerns for excessive documentation and monitoring. 1 * 2 mL
Catalog # GENO786-709
Supplier:  G BIOSCIENCES
Our No-Waste format adjuvant minimizes waste and risk of cross contamination. Small dose packaging allows researchers to assign a vial per project or animal without concerns for excessive documentation and monitoring. 1 * 2 mL
Catalog # GENO786-709
Supplier:  G BIOSCIENCES
Frequently Bought Together