Order Entry
Slovakia
ContactUsLinkComponent
An ELISA kit for the detection of TNFRSF13C/BAFFR (Human). This uses Sandwich ELISA, Double Antibody and has a sensitivity of 0.094n g/ml. 1 * 1 KIT
Catalog # GENOIT1746
Supplier:  G BIOSCIENCES
An ELISA kit for the detection of TNFRSF13C/BAFFR (Human). This uses Sandwich ELISA, Double Antibody and has a sensitivity of 0.094n g/ml. 1 * 1 KIT
Catalog # GENOIT1746
Supplier:  G BIOSCIENCES
Frequently Bought Together