Order Entry
Slovakia
ContactUsLinkComponent
An ELISA kit for the detection of UBE2A (Human). This uses Sandwich ELISA, Double Antibody and has a sensitivity of 0.188ng/ml. 1 * 1 KIT
Catalog # GENOIT2336
Supplier:  G BIOSCIENCES
An ELISA kit for the detection of UBE2A (Human). This uses Sandwich ELISA, Double Antibody and has a sensitivity of 0.188ng/ml. 1 * 1 KIT
Catalog # GENOIT2336
Supplier:  G BIOSCIENCES
Frequently Bought Together