Order Entry
Slovakia
ContactUsLinkComponent
An ELISA kit for the detection of TF/F3 (Human). This uses Sandwich ELISA, Double Antibody and has a sensitivity of 4.688pg/ml. 1 * 1 KIT
Catalog # GENOIT1738
Supplier:  G BIOSCIENCES
An ELISA kit for the detection of TF/F3 (Human). This uses Sandwich ELISA, Double Antibody and has a sensitivity of 4.688pg/ml. 1 * 1 KIT
Catalog # GENOIT1738
Supplier:  G BIOSCIENCES
Frequently Bought Together