Order Entry
Slovakia
ContactUsLinkComponent
An ELISA kit for the detection of IL-10 (Human). This uses Sandwich ELISA, Double Antibody and has a sensitivity of 4.688pg/ml. 1 * 1 KIT
Catalog # GENOIT1634
Supplier:  G BIOSCIENCES
An ELISA kit for the detection of IL-10 (Human). This uses Sandwich ELISA, Double Antibody and has a sensitivity of 4.688pg/ml. 1 * 1 KIT
Catalog # GENOIT1634
Supplier:  G BIOSCIENCES
Frequently Bought Together