Order Entry
Slovakia
ContactUsLinkComponent
G-Sep is a gel filtration resin comprised of ultrapure cross-linked dextran for desalting and buffer exchange in industrial applic ations. It exhibits high selectivity, superb resolution, low non-specific adsorption and robust chemical stability. null
Katalógové číslo GENO786-1403
Supplier:  G BIOSCIENCES
G-Sep is a gel filtration resin comprised of ultrapure cross-linked dextran for desalting and buffer exchange in industrial applic ations. It exhibits high selectivity, superb resolution, low non-specific adsorption and robust chemical stability. null
Katalógové číslo GENO786-1403
Supplier:  G BIOSCIENCES
Frequently Bought Together