NEEDLE HOLDER 58MM LUER-LOCK FOR 860 1 * 1 items
Catalog # METR6.2049.040
Supplier: METROHM
Frequently Bought Together
[EN]FRITTE Ø12mm, SET 1 * 50 ST
Supplier: BUCHI
mains filter max.1x250V FN2010-3-06 1 * 1 items
Supplier: NABERTHERM
GASKET KIT CryoPlus 1 SRO ONLY 1 * 1 items
Supplier: Thermo Fisher Scientific
Handlebar for inner glass door (Right) UN UF IN IF 1 * 1 items
Supplier: MEMMERT
Baseboard length: 434,5 mm ICO 105, 150 1 * 1 items
Supplier: MEMMERT
Tube socket 75/85, Mat 1.4301 Flange diam 55,Tube diam 26,9x2,0mm 1 * 1 items
Supplier: NABERTHERM
DRIVE COMPLETE GM 300 1 * 1 items
Supplier: RETSCH
Polarimeter tubes and spare parts
Supplier: BELLINGHAM STANLEY
GAS BALLAST TUBE 56,4 MM LONG 1 * 1 items
Supplier: VACUUBRAND
Cup seal 1 * 1 items
Supplier: Heidolph Instruments GmbH & Co.KG
Cylinder isolation kpl 1 * 1 items
Supplier: BINDER GMBH
BIOTECTOR ACID REAGENT WITH CATALYST 1 * 20 L
Supplier: HACH
cable lug 1 * 1 items
Supplier: ELMA SCHMIDBAUER
HEATING ELEMENT 1 * 1 items
Supplier: VWR Collection
Pocket Pro Plus Tester sample cup/c 1 * 1 items
Supplier: HACH