Order Entry
Slovakia
ContactUsLinkComponent
Stericlin® sterilisation wrap sheet, non-woven 59g/m² blue, 60x60cm, DIN EN ISO 11607 1 * 500 items
Catalog # 129-1388
Stericlin® sterilisation wrap sheet, non-woven 59g/m² blue, 60x60cm, DIN EN ISO 11607 1 * 500 items
Catalog # 129-1388
Frequently Bought Together