Order Entry
Slovakia
ContactUsLinkComponent
Stericlin® sterilisation wrap sheet, SMMS 60g/m² dark blue, 140x140cm, DIN EN ISO 11607 1 * 80 items
  129-1369
Stericlin® sterilisation wrap sheet, SMMS 60g/m² dark blue, 140x140cm, DIN EN ISO 11607 1 * 80 items
  129-1369
Frequently Bought Together