Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovakia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
20 ml Precision FAST qPCR Master Mix with ROX at a reduced level premixed with SYBRgreen (20x 1ml) 1 * 2.000 Tests
Catalog # ZPFASTLRSY20ML
Supplier:
PRIMERDESIGN
Expand image
20 ml Precision FAST qPCR Master Mix with ROX at a reduced level premixed with SYBRgreen (20x 1ml) 1 * 2.000 Tests
Catalog # ZPFASTLRSY20ML
Supplier:
PRIMERDESIGN
Frequently Bought Together
Restriction enzyme FastDigest Eco147I 100µl (100 reactions) 1 * 1 KIT
Supplier: Thermo Fisher Scientific
Buffer pack PCR Phusion GC final 1x buffer concentration equals 1,5mM MgCl2, contains 4x1,5ml of 5x buffer, 2x1,5ml of 50mM MgCl2 so lution, 1x0,5ml of 100% DMSO 1 * 1 KIT
Supplier: FINNZYMES REAGENTS