Order Entry
Slovakia
ContactUsLinkComponent
Sweet wort hydrometer, 15-18:0,05%mas, with therm. +5+30:0,5C with red or blue special liquid, with correction scale, reading below , ref. temp. 20C, 400mm long 1 * 1 items
Catalog # AMARH847368-FL
Supplier:  Amarell
Sweet wort hydrometer, 15-18:0,05%mas, with therm. +5+30:0,5C with red or blue special liquid, with correction scale, reading below , ref. temp. 20C, 400mm long 1 * 1 items
Catalog # AMARH847368-FL
Supplier:  Amarell
Frequently Bought Together