Order Entry
Slovakia
Orders LinkContactUsLinkComponent
 

Human recombinant CD99

10 µG
  APOSBITP1511-10UG
Human recombinant CD99
  APOSBITP1511-10UG
 :  
Human recombinant CD99

10 µG

 :  
 
263,05 €
   
1 * 10 µG
 
 

 

  • Pk:
    10 µG
  • Cat. No.:
    APOSBITP1511-10UG
  • Protein/peptide type:
    Recombinant
  • Protein/peptide name:
    CD99
  • Species:
    Human