Order Entry
Slovakia
Orders LinkContactUsLinkComponent
 

2-Hydroxy-1-naphthaldehyde

Bottle, 500 g
Catalog # APOSOR8817-500G
Irritant
Environment
Supplier:   Apollo Scientific
2-Hydroxy-1-naphthaldehyde
Catalog # APOSOR8817-500G
Supplier:  Apollo Scientific
CAS Number:  708-06-5
2-Hydroxy-1-naphthaldehyde

Bottle, 500 g

CAS Number:  708-06-5
Price
190,71 €
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  2-Hydroxy-α-naphthaldehyde
  • CAS Number:708-06-5
  • Formula:C₁₁H₈O₂
  • MDL Number:MFCD00004005
  • Molecular Weight:172.18 g/mol
  • Boiling Pt:191…193 °C/27mmHg
  • Melting Pt:76…80 °C
  • Storage Temperature:Ambient
  • EINECS:211-902-0
  • UN:3077
  • ADR:9+EHS,III
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 g

Specifications