HT-2 Toxin standard solution
HT-2 Toxin, 100 µg/ml in acetonitrile, Bottle, 5 mL
APOSBIMS115-5ML
Analytical reference standard solution in acetonitrile.
: Expires in 12 months.
- Concentration:100 µg/ml in acetonitrile
- Pack type:Bottle
- Pk:5 mL
- Description:HT-2 Toxin
- Cat. No.:APOSBIMS115-5ML
- Formula:C₂₂H₃₂O₈
- Description:HT-2 Toxin
- Storage temperature:Freezer
- Flash point:2 °C
- CAS No.:26934-87-2



