Ochratoxin A standard solution
Ochratoxin A, 10 µg/ml in acetonitrile, Bottle, 1 mL
Dodavatel: Apollo Scientific
Ochratoxin A standard solution
Katalogové číslo APOSBIMS107-1ML
Dodavatel: Apollo Scientific
Číslo CAS:
Přejít na:
Product Details & Documents
Analytical reference standard solution in acetonitrile.
Caution: Expires in 12 months.
Documents
Certifikáty
Hledat podle čísla šaržeSpecifikace
Specifikace
- Concentration:10 µg/ml in acetonitrile
- Pack type:Bottle
- Pk:1 mL
- Description:Ochratoxin A
- Cat. No.:APOSBIMS107-1ML
- Formula:C₂₀H₁₈ClNO₆
- Description:Ochratoxin A
- Storage temperature:Freezer
- Flash point:2 °C
- CAS No.:303-47-9
Specifikace
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



