Order Entry
Slovakia
ContactUsLinkComponent
Pcr tube, 0.5ml, flatcap, yellow, case of 1000. the thin wall construction optimises the heat transfer from the block to the reactio n solution. 1 * 1.000 items
Catalog # GREI682276_1000
Supplier:  Greiner Bio-One
Pcr tube, 0.5ml, flatcap, yellow, case of 1000. the thin wall construction optimises the heat transfer from the block to the reactio n solution. 1 * 1.000 items
Catalog # GREI682276_1000
Supplier:  Greiner Bio-One
Frequently Bought Together