Order Entry
Slovakia
ContactUsLinkComponent
Blood grouping tubes are available in two formulations, acd-a, acd-b or with a cpda solution. blood grouping tubes are for blood gro uping determinations and also for cell preservation. 1 * 1.200 items
Catalog # GREI455056_1200
Supplier:  Greiner Bio-One
Blood grouping tubes are available in two formulations, acd-a, acd-b or with a cpda solution. blood grouping tubes are for blood gro uping determinations and also for cell preservation. 1 * 1.200 items
Catalog # GREI455056_1200
Supplier:  Greiner Bio-One
Frequently Bought Together