ICH/USP oral target elements standard C, Au, Ir, Os, Pd, Pt, Rh, and Ru 100 µg/mL in 15 % HCl 1 * 1 items
Catalog # HEWL5190-9768
Supplier: AGILENT HEWLETT PACKARD
ICH/USP oral target elements standard C, Au, Ir, Os, Pd, Pt, Rh, and Ru 100 µg/mL in 15 % HCl 1 * 1 items
Catalog # HEWL5190-9768
Supplier: AGILENT HEWLETT PACKARD
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Sealing film for manual application, Parafilm® M
Supplier: HEATHROW
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR® Facial Tissues
Supplier: VWR Collection